Cleanroom tapes, PCX™
Toimittaja: MICRONOVA
PCX-1RE PCX-2RE PCX-2DB PCX-1YE PCX-2YE PCX-1WH_U PCX-34RE PCX-34WH PCX-2OR PCX-1TR PCX-1OR PCX-3OR PCX-2DG PCX-1BL PCX-1DG PCX-2WH PCX-34WHIR PCX-1DB PCX-1WH
MICRPCX-1REEA 1800 EUR
MICRPCX-1RE MICRPCX-2RE MICRPCX-2DB MICRPCX-1YE MICRPCX-2YE 819-0031 MICRPCX-34RE MICRPCX-34WH MICRPCX-2OR MICRPCX-1TR MICRPCX-1OR MICRPCX-3OR MICRPCX-2DG MICRPCX-1BL MICRPCX-1DG MICRPCX-2WH MICRPCX-34WHIR MICRPCX-1DB MICRPCX-1WH
Cleanroom tapes, PCX™
Teipit
These cleanroom tapes are made of LDPE with acrylic adhesive.
  • Labeling
  • Sealing, wafer box sealing
  • Over labeling (protect paper labels in water baths)
A low density polyethylene tape, low in halogens and leachable chlorides. Processed and packaged in a cleanroom. The polyethylene film and acrylic adhesive lessen the chance of outgassing. PCX offers excellent chemical resistance and good conformability. Works well with polycarbonates, glass and acrylics. Suitable for the following:
Length: 32,9 m
Different colours available.