Flat mops
Toimittaja: MICRONOVA
MZFM7-14 MZFM7-14IR MZFM7-18 MZFM7-18IR MZFM7-9
MICRMZFM7-14EA 38.5 EUR
MICRMZFM7-14 MICRMZFM7-14IR MICRMZFM7-18 MICRMZFM7-18IR MICRMZFM7-9
Flat mops
Siivousvälineet
These brick-style, flat sponge mop heads are excellent for applying disinfectant in pharmaceutical production or aseptic processing applications.
  • Easily clean large surfaces
  • Heavy-duty construction offers additional resistance to textured floors and aggressive cleaning methods
  • Fully padded mop heads can be laundered and reused several times
Ideal for cleaning large surface areas as well as reaching shelving, doorjambs, and corners.
Mop heads feature heavy-duty construction with PolyTex™ textured polyester material, PolyTex™ material combined withMore Product Information