Masterflex® Particle Tolerant Solenoid Valves, Avantor®
Toimittaja: Avantor Fluid Handling
98305-26 98305-34 98305-42 98305-24 98305-32 98305-36
MFLX98305-26EA 225 EUR
MFLX98305-26 MFLX98305-34 MFLX98305-42 MFLX98305-24 MFLX98305-32 MFLX98305-36
Masterflex® Particle Tolerant Solenoid Valves, Avantor®
Venttiilit Solenoidi venttiilit
Particles will not cause damage.
  • Special elastomer diaphragm and seals are forgiving of particulate matter that would typically damage a plastic valve seat
  • Inert materials — excellent chemical resistance
  • Molded PEEK valve bodies
  • Excellent choice for demanding low-flow applications where minimum size and internal volume are important
These Masterflex® particle tolerant solenoid valves feature a apecial Perfluor™ soft elastomer diaphragm and seals, which are forgiving of particulate matter that would typically damage a plastic valve seat. Valves areMore Product Information