Masterflex® Solenoid-Operated Pinch Valves for Metric Tubing, Avantor®
Toimittaja: Avantor Fluid Handling
98305-00 98305-02 98305-03 98305-05 98305-10 98305-11 98305-12 98305-13 98305-15 98305-01 98305-06 98305-07 98305-14
MFLX98305-00EA 225 EUR
MFLX98305-00 MFLX98305-02 MFLX98305-03 MFLX98305-05 MFLX98305-10 MFLX98305-11 MFLX98305-12 MFLX98305-13 MFLX98305-15 MFLX98305-01 MFLX98305-06 MFLX98305-07 MFLX98305-14
Masterflex® Solenoid-Operated Pinch Valves for Metric Tubing, Avantor®
Venttiilit Solenoidi venttiilit
Excellent choice for viscous fluids.
  • Specifically designed for metric sized tubing
These solenoid-operated 2- and 3-Way pinch valves are ideal for use with fluids that are difficult for traditional solenoid valves to handle, such as viscous fluids. They are ideal for on/off flow control applications and where frequent changes of wetted parts are necessary. All models are compatible with silicone, C-FLEX®, and PharMed® BPT tubing. The two-way valves are available in both normally closed and normally open configurations. The three-way valves accept twoMore Product Information