Dynalon Two-Way Stopcocks with Hose Barb Connections
Toimittaja: Avantor Fluid Handling
98261-10 98261-20 98261-30
MFLX98261-10EA 8.5 EUR
MFLX98261-10 MFLX98261-20 MFLX98261-30
Dynalon Two-Way Stopcocks with Hose Barb Connections
Hanat
Easily adjust flow.
  • Miniature plastic stopcocks have a straight bore
  • Ideal for liquids and gases under low pressure (operating pressure 7.25 psi/0.5 bar) or low vacuum.
The low friction properties of high density polyethylene in these miniature plastic stopcocks allows for easy turning of PP key which requires no lubricant, thus eliminating blockage of the bores with grease.