Evsco Valves Manual and Actuated Accessories
Toimittaja: Avantor Fluid Handling
98150-50 98150-51
MFLX98150-50EA 25.6 EUR
MFLX98150-50 MFLX98150-51
Evsco Valves Manual and Actuated Accessories
Venttiilit Elliptiset venttiilit
Safely mount your plastic elliptical valves.
  • Optimize performance of your Evsco Valves
Constucted of durable polypropylene, these mounting brackets are ideal for making your valve more versitile. These mounting brackets allow for wall, panel, or undermount.