Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, FDA-Compliant Viton®, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX97629-26EA 206 EUR
MFLX97629-26 MFLX97629-42 MFLX97629-18 MFLX97629-22 MFLX97629-24 MFLX97629-28 MFLX97629-32 MFLX97629-34 MFLX97629-36 MFLX97629-40 MFLX97629-44 MFLX97629-30 MFLX97629-38 MFLX97629-46 MFLX97629-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, FDA-Compliant Viton®, Avantor®
Letku Pumppuletku
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Perfect for food and lab applications where FDA compliance is required
  • Excellent chemical compatibility
To complete your Ismatec® pumping system, order three-stop color-coded tubing andMore Product Information