Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® LMT-55, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX97619-09EA 102 EUR
MFLX97619-09 MFLX97619-10 MFLX97619-12 MFLX97619-14 MFLX97619-16 MFLX97619-18 MFLX97619-19 MFLX97619-22 MFLX97619-24 MFLX97619-26 MFLX97619-27 MFLX97619-28 MFLX97619-29 MFLX97619-30 MFLX97619-31 MFLX97619-32 MFLX97619-34 MFLX97619-36 MFLX97619-38 MFLX97619-39 MFLX97619-40 MFLX97619-42 MFLX97619-44 MFLX97619-46 MFLX97619-48 MFLX97619-49
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® LMT-55, Avantor®
Letku Pumppuletku
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attachedMore Product Information