Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC, Avantor®
Toimittaja: Avantor Fluid Handling
96605-20 96605-22 96605-24 96605-27 96605-29 96605-31 96605-33 96605-37 96605-39 96605-41 96605-35 96605-43
MFLX96605-20EA 45.9 EUR
MFLX96605-20 MFLX96605-22 MFLX96605-24 MFLX96605-27 MFLX96605-29 MFLX96605-31 MFLX96605-33 MFLX96605-37 MFLX96605-39 MFLX96605-41 MFLX96605-35 MFLX96605-43
Masterflex® Transfer Tubing, Cleargreen® Phthalate and DEHP-Free PVC, Avantor®
Letku Siirtoputki
Eliminate odor transferring to sample.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Characteristics: Translucent, odorless, low water absorption, low extractables
Certified compliant FDA compliant, USP Class VI, 3A compliant, Reach compliant and RoHS compliant.
Temperature range: -58 to 165 °F (-50 to 74 °C)More Product Information