Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-LFL, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX96450-10EA 165 EUR
MFLX96450-10 MFLX96450-12 MFLX96450-18 MFLX96450-26 MFLX96450-28 MFLX96450-30 MFLX96450-33 MFLX96450-34 MFLX96450-42 MFLX96450-43 MFLX96450-48 MFLX96450-14 MFLX96450-22 MFLX96450-24 MFLX96450-36 MFLX96450-37 MFLX96450-40
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Tygon® S3™ E-LFL, Avantor®
Letku Pumppuletku
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Longest pumping life of any Tygon® formulation
  • Extremely low particulate spallation
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. TubingMore Product Information