Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-LFL, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX96449-10EA 159 EUR
MFLX96449-10 MFLX96449-12 MFLX96449-14 MFLX96449-18 MFLX96449-26 MFLX96449-30 MFLX96449-33 MFLX96449-34 MFLX96449-36 MFLX96449-37 MFLX96449-42 MFLX96449-43 MFLX96449-48 MFLX96449-22 MFLX96449-24 MFLX96449-28 MFLX96449-40 MFLX96449-31
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-LFL, Avantor®
Letku Pumppuletku
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Longest tubing life of all Tygon tubing formulations
  • Broad chemical compatibility
  • Clear for easy flow monitoring
To complete your Ismatec® pumping system, order two-stop color-coded andMore Product Information