Masterflex® L/S® Precision Pump Tubing, Puri-Flex®, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX96419-14EA 70 EUR
MFLX96419-14 MFLX96419-13 MFLX96419-16 MFLX96419-17 MFLX96419-18 MFLX96419-25
Masterflex® L/S® Precision Pump Tubing, Puri-Flex®, Avantor®
Letku Pumppuletku
Long life tubing for any Masterflex® pump used in your pharma and bioprocess applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options