Masterflex® I/P® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX96412-26EA 896 EUR
MFLX96412-26 MFLX96412-73
Masterflex® I/P® Precision Pump Tubing, FDA-Compliant Viton®, Avantor®
Letku Pumppuletku
Safely pump harsh chemicals.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory Approvals to match your unique application
Masterflex® FDA-Compliant Viton® Precision pump tubing is designed for enhancedMore Product Information