Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX96410-13EA 87.7 EUR
MFLX96410-13 MFLX96410-14 MFLX96410-16 MFLX96410-17 MFLX96410-18 MFLX96410-25
Masterflex® L/S® Precision Pump Tubing, Platinum-Cured Silicone, Avantor®
Letku Pumppuletku
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and