Masterflex® L/S® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX96403-13EA 1520 EUR
MFLX96403-13 MFLX96403-14 MFLX96403-15 MFLX96403-16 MFLX96403-17 MFLX96403-18 MFLX96403-24 MFLX96403-25 MFLX96403-35 MFLX96403-36
Masterflex® L/S® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Letku Pumppuletku
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and