Masterflex® L/S® Pump Tubing, Gore® STA-PURE® PFL-Series, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX96242-14EA 1220 EUR
MFLX96242-14 MFLX96242-15 MFLX96242-16 MFLX96242-17 MFLX96242-18 MFLX96242-24 MFLX96242-25 MFLX96242-35 MFLX96242-36
Masterflex® L/S® Pump Tubing, Gore® STA-PURE® PFL-Series, Avantor®
Letku Pumppuletku
High-performance pump tubing for challenging applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Custom extruded to fit Masterflex® pumps
  • Lot-to-lot