Sani-Tech® Ultra Platinum-Cured Silicone Lab Tubing
Toimittaja: Avantor Fluid Handling
96201-63
MFLX96201-19EA 57.6 EUR
MFLX96201-19 MFLX96201-50 MFLX96201-55 MFLX96201-60 MFLX96201-63
Sani-Tech® Ultra Platinum-Cured Silicone Lab Tubing
Letku Siirtoputki
Don't let extractables ruin your formulations.
  • Exceptionally low TOCs minimize fluid contamination
  • Ultra-pure silicone will not impart flavors or odors
  • Superior pumping life reduces downtime for tubing changes
Applications: Sterile fill and transfer, cell harvest, media processing, sanitary peristaltic pumping, production filtration and fermentation, bioreactor process lines, and pharmaceutical delivery.
Characteristics: Excellent flexibility, long pump life, non-yellowing, tasteless and odorless, nontoxic, translucent.
Certifications: Elastomer is FDA-More Product Information