Masterflex® Transfer Tubing, Black Rubber Vacuum, Avantor®
Toimittaja: Avantor Fluid Handling
96143-03 96143-05 96143-07 96143-09 96143-11 96143-15 96143-17 96143-13
MFLX96143-03EA 172 EUR
MFLX96143-03 MFLX96143-05 MFLX96143-07 MFLX96143-09 MFLX96143-11 MFLX96143-15 MFLX96143-17 MFLX96143-13
Masterflex® Transfer Tubing, Black Rubber Vacuum, Avantor®
Letku Siirtoputki
Suitable for vacuum use.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
The tubing is ideal for vacuum and pressure applications for general lab and industrial use. It features thick flexible walls with a temperature range of 0 to 150 °F (-18 to 65 °C). Can be sterilized with ethylene oxide or autoclave for 30More Product Information