Dow Corning® Pharma-50 Transfer Tubing
Toimittaja: Avantor Fluid Handling
MFLX96105-00EA 136 EUR
MFLX96105-00 MFLX96105-03 MFLX96105-04 MFLX96105-05 MFLX96105-08 MFLX96105-12 MFLX96105-02 MFLX96105-09 MFLX96105-11
Dow Corning® Pharma-50 Transfer Tubing
Letku Siirtoputki
Pharma tubing remains flexible and resilient for long service life.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Pharmaceutical, biotechnology , and ultra-pure fluid processing.
Characteristics: 50 durometer, translucent platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Pharma tubing remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silica to siliconeMore Product Information