Masterflex® Transfer Tubing, Premo Flex®, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX95953-07EA 517 EUR
MFLX95953-07 MFLX95953-01 MFLX95953-03 MFLX95953-05
Masterflex® Transfer Tubing, Premo Flex®, Avantor®
Letku Siirtoputki
Resists weather and ozone.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Outstanding resistance to heat and ozone
Masterflex® Premp Flex® transfer tubing is ideal for use with fuels such as petroleum oils, gasoline, kerosene, and lubricating oils, while maintaining tube integrity. The Buna-N tube, modified Buna-N nitrile tube cover, along with the high tensile textile core tube reinforcement makes this tubing suitable for high pressureMore Product Information