Thermo Scientific™ Nalgene™ 489 Linear LDPE Tubing
Toimittaja: Avantor Fluid Handling
95912-01 95912-03 95912-04 95912-06
MFLX95912-01EA 101 EUR
MFLX95912-01 MFLX95912-03 MFLX95912-04 MFLX95912-06
Thermo Scientific™ Nalgene™ 489 Linear LDPE Tubing
Letku Siirtoputki
Suitable for food processing applications.
  • Global market leader in range of formulations for most extensive chemical compatibility
Thermo Scientific Nalgene 489 linear low-density polyethylene (LLDPE) is a general purpose tubing suitable for many applications. Made of FDA compliant resin, is it suitable for food processing applications as well as general laboratory or industrial use. 489 LLDPE offers broad chemical resistance and is more flexible than other polyethylene formulations. This translucent formulation is nontoxic, odorless, and tasteless.More Product Information