Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, PharMed® BPT, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX95714-10EA 140 EUR
MFLX95714-10 MFLX95714-12 MFLX95714-14 MFLX95714-18 MFLX95714-22 MFLX95714-24 MFLX95714-26 MFLX95714-28 MFLX95714-30 MFLX95714-32 MFLX95714-34 MFLX95714-36 MFLX95714-38 MFLX95714-40 MFLX95714-42 MFLX95714-44 MFLX95714-46 MFLX95714-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, PharMed® BPT, Avantor®
Letku Pumppuletku
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Recommended for cell cultures and tissues
  • Impermeable to normal light and UV-radiation
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. TubingMore Product Information