Masterflex® Transfer Tubing, Tygon® SE-200, Non-DEHP FEP-Lined, Avantor®

Toimittaja: Avantor Fluid Handling
95713-00 95713-01 95713-10 95713-30 95713-41 95713-50 95713-20 95713-60
MFLX95713-00EA 196 EUR
MFLX95713-00 MFLX95713-01 MFLX95713-10 MFLX95713-30 MFLX95713-41 MFLX95713-50 MFLX95713-20 MFLX95713-60
Masterflex® Transfer Tubing, Tygon® SE-200, Non-DEHP FEP-Lined, Avantor®
Letku Siirtoputki
Keep samples free of odors and taste.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application

Formulation: Tygon® SE-200
Applications: Chemical and pharmaceutical processing, beverage dispensing, paint and solvent production, semiconductor processing
Characteristics: Bio-based, phthalate-free. Flexible, chemicallyMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR