Masterflex® Ismatec® Microbore Pump Tubing, Solva™ Solvent/Hydrocarbon, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX95712-12EA 78.6 EUR
MFLX95712-12 MFLX95712-14 MFLX95712-18 MFLX95712-26 MFLX95712-28 MFLX95712-30 MFLX95712-32 MFLX95712-34 MFLX95712-36 MFLX95712-40 MFLX95712-42 MFLX95712-44 MFLX95712-48 MFLX95712-10 MFLX95712-22 MFLX95712-24 MFLX95712-38
Masterflex® Ismatec® Microbore Pump Tubing, Solva™ Solvent/Hydrocarbon, Avantor®
Letku Pumppuletku
Specifically designed for use with hydrocarbons, petroleum products and distillates.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
  • Resists most hydrocarbons, embrittlement, and swelling
Resistant to ozone and UV light — won't crack, swell, or harden.