Masterflex® Transfer Tubing, Tygon® ND-100-65, Non-DEHP, Avantor®

Toimittaja: Avantor Fluid Handling
95666-00 95666-04 95666-05 95666-06 95666-07 95666-08 95666-09 95666-10 95666-11 95666-13 95666-15 95666-16 95666-17 95666-20 95666-21 95666-22 95666-26 95666-27 95666-28 95666-31 95666-01 95666-03 95666-14 95666-12 95666-33
MFLX95666-00EA 52.5 EUR
MFLX95666-00 MFLX95666-04 MFLX95666-05 MFLX95666-06 MFLX95666-07 MFLX95666-08 MFLX95666-09 MFLX95666-10 MFLX95666-11 MFLX95666-13 MFLX95666-15 MFLX95666-16 MFLX95666-17 MFLX95666-20 MFLX95666-21 MFLX95666-22 MFLX95666-26 MFLX95666-27 MFLX95666-28 MFLX95666-31 MFLX95666-01 MFLX95666-03 MFLX95666-14 MFLX95666-12 MFLX95666-33
Masterflex® Transfer Tubing, Tygon® ND-100-65, Non-DEHP, Avantor®
Letku Siirtoputki
Drains completely.

  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application

Formulation: ND-100-65
Applications: Pharmaceutical and biotech research applications.
Characteristics:Non-DEHP, phthalate free. Non-wetted surface permits complete drainage. Bondable, weldable. Clear.
Certification: USP Class VI, REACH, ISO 10993
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR