Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Toimittaja: Avantor Fluid Handling
95505-00 95505-02 95505-04 95505-06 95505-14 95505-18 95505-20 95505-22 95505-24 95505-42 95505-08 95505-16 95505-26 95505-28 95505-44
MFLX95505-00EA 36 EUR
MFLX95505-00 MFLX95505-02 MFLX95505-04 MFLX95505-06 MFLX95505-14 MFLX95505-18 MFLX95505-20 MFLX95505-22 MFLX95505-24 MFLX95505-42 MFLX95505-08 MFLX95505-16 MFLX95505-26 MFLX95505-28 MFLX95505-44
Masterflex® Transfer Tubing, DEHP-Free Flexible PVC, Avantor®
Letku Siirtoputki
As flexible as you need it.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Analytical instruments, general laboratory. Use for transfer of liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications.
Characteristics:Nontoxic DEHP-free formulation.More Product Information