Hubtron™ Black Advance Fluoroelastomer Tubing
Toimittaja: Avantor Fluid Handling
95256-00
MFLX95256-02EA 765 EUR
MFLX95256-02 MFLX95256-04 MFLX95256-06 MFLX95256-00
Hubtron™ Black Advance Fluoroelastomer Tubing
Letku Siirtoputki
Ideal for corrosive chemical transfer.
  • Global market leader in range of formulations for most extensive chemical compatibility
Applications: Corrosive chemical transfer, electronics applications, ink and printing, and general manufacturing applications requiring UV protection.|Characteristics: Opaque black with UV block, very low gas permeability, highly chemically compatible. Inner fluoroplastic liner, outer fluoroelastomer cover. Excellent chemical resistance to acids, alkalis, alcohols, and esters.
Certification: None.
Temperature: 14 to 302 °F (–5More Product Information