Masterflex® Transfer Tubing Spool, Fluorinated Ethylene Propylene (FEP), Avantor®
Toimittaja: Avantor Fluid Handling
95230-02 95230-08 95230-10 95230-12 95230-14
MFLX95230-00EA 422 EUR
MFLX95230-00 MFLX95230-04 MFLX95230-06 MFLX95230-02 MFLX95230-08 MFLX95230-10 MFLX95230-12 MFLX95230-14
Masterflex® Transfer Tubing Spool, Fluorinated Ethylene Propylene (FEP), Avantor®
Letku Siirtoputki
Maximize tubing on hand.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
This tubing is ideal for general laboratory use. They offer long life, won't age or oxidize, fits tightly to glass and metal, bends to small radius curves, are nontoxic. FDA compliant and made according to GMP. Temperature range is -51 toMore Product Information