Masterflex® L/S® Digital Process Pump Systems with Cytoflow™ Pump Head, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX77975-50-EUEA 8160 EUR
MFLX77975-50-EU MFLX77975-52-EU
Masterflex® L/S® Digital Process Pump Systems with Cytoflow™ Pump Head, Avantor®
Pumput Peristalttiset pumput
Ideal for the transfer and dispensing of live cell cultures, media, and other shear-sensitive fluids in process and washdown environments.
  • Convex-roller design is gentler on live cells and shear-sensitive fluids—validated by independent test data
  • Large-diameter rotor delivers higher flow rates at lower speeds
  • Low-speed pumping lets you maintain the consistency and characteristics of shear-sensitive fluids
  • Drive housings are completely sealed—IP66 and NEMA 4X rated—simply hose down to clean
  • Brushless, maintenance-free motor offers ±0.1% speed control