Masterflex® L/S® Pump Head Adapters, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX77495-00EA 1100 EUR
MFLX77495-00 MFLX77495-20
Masterflex® L/S® Pump Head Adapters, Avantor®
Pumput Pumppujen käyttölaitteet Peristalttiset pumppukäytöt
Customize your pump—mount L/S® pump heads to a 56C frame motor.
  • Use up to 2 L/S® pump heads—pump two fluids simultaneously at equal or different flow rates
  • IP56-rated cast-aluminum adapters feature built-in gear box
These adapters easily mount Masterflex® L/S® pump heads to a 56C frame motor (pump head and motor sold separately). Adapters accept seven different pump head types: Standard, Easy-Load®, Easy-Load ® II, High-Performance, Cartridge, PTFE-Tubing, and PTFE-Diaphragm
Use the following formula to calculate flow rate— do not exceed 600 rpm pumpMore Product Information