Masterflex® I/P® Digital Network-Compatible Pump Systems
Toimittaja: Avantor Fluid Handling
MFLX77420-82-EUEA 11830 EUR
MFLX77420-82-EU
Masterflex® I/P® Digital Network-Compatible Pump Systems
Pumput Peristalttiset pumput
Networking drives offer either native EtherNet/IP™ or Profibus communication-no additional hardware or firmware required.
  • Flow rate 0.0006 to 26 LPM with I/P tubing-flow rate depends on drive rpm, pump head, number of pump heads, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mod- continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication-voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and