Masterflex® L/S® Easy-Load® II Pump Head Tubing Retainer Kits, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX77200-90EA 82.4 EUR
MFLX77200-90 MFLX77200-92 MFLX77200-94
Masterflex® L/S® Easy-Load® II Pump Head Tubing Retainer Kits, Avantor®
Pumput Peristalttiset pumput
Enhance the life of your pump head with the replacement retainer kits.
  • Retainer kits provide the parts necessary to replace the spring-loaded tubing retainers in your Masterflex® Easy-Load® II pump heads
Replacement parts to keep your pump head operating even longer. Tubing retainers hold the tubing in place within the pump head. Pump head sold separately.
Toimitustiedot: Includes two small upper retainers, two small lower retainers, and two compression springs.