Masterflex® L/S® Compact Variable-Speed Drive, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX77200-12EA 962 EUR
MFLX77200-12 MFLX77200-20-EU
Masterflex® L/S® Compact Variable-Speed Drive, Avantor®
Pumput Pumppujen käyttölaitteet Peristalttiset pumppukäytöt
Stackable, compact pump drive delivers versatile performance in limited space.
  • Flow rate 2.1 to 560 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Stackable, compact drive is perfect for small work areas
  • Constructed with a painted steel housing with IP22 rating
  • No difficult programming—single switch for forward/off/reverse and analog controller
  • Pump in either direction—also allows purging before or after pumping
  • Remote control of start/stop via contact closure
These compact drives are a perfect when space is limited, as they fitMore Product Information