Stainless Steel Motor Sanitary Centrifugal Pumps
Toimittaja: Avantor Fluid Handling
76720-05 76720-20 76720-40
MFLX76720-05EA 3150 EUR
MFLX76720-05 MFLX76720-20 MFLX76720-40
Stainless Steel Motor Sanitary Centrifugal Pumps
Pumput Keskipakopumput
Clean-in-place (CIP) 3A sanitary transfer pump.
  • 316L stainless steel liquid end construction
  • Washdown motors available in stainless steel
  • Stainless steel adapters are standard
  • Ideal for RO water and pharmaceutical manufacturing
Piping changes are eliminated due to the interchangeable design that these economical pumps offer when compared to other pumps. Models are available with inlets up to 6" contact our Application Specialists for assistance in sizing the proper pump for your system.
Toimitustiedot: Includes conduit box for hardwiring.