Masterflex® Single-Use Forceps, Avantor®
Toimittaja: Avantor Fluid Handling
75955-11 75955-13 75955-15
MFLX75955-11EA 154 EUR
MFLX75955-11 MFLX75955-13 MFLX75955-15
Masterflex® Single-Use Forceps, Avantor®
Pinsetit
No more cross-contamination.
  • Protect yourself and your samples with disposable forceps
  • Autoclavable plastic or stainless steel construction
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.