Masterflex® Entry-Level Touch-Screen Syringe Pumps, Avantor®
Toimittaja: Avantor Fluid Handling
74905-08 74905-12 74905-34
MFLX74905-02EA 2740 EUR
MFLX74905-02 MFLX74905-04 MFLX74905-06 MFLX74905-08 MFLX74905-12 MFLX74905-34
Masterflex® Entry-Level Touch-Screen Syringe Pumps, Avantor®
Pumput Ruiskupumput
Accommodate up to two syringes in a cost-effective touch-screen syringe pump.
  • Flow ranges from 0.54 pL up to 88.8 ml/min
  • Full-color touch screen
  • Daisy chain multiple pumps using the integrated RS-485
  • Use with any manufacturer syringe—glass, stainless steel, or plastic—from 0.5 ìL to 60 mL
These entry-level pumps offer features normally found in more expensive models. An intuitive touch-screen display lets you see all operating parameters and easily create and recall configurations. Front-panel LED light indicates when pump is operating—vie