Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Toimittaja: Avantor Fluid Handling
70607-00 70607-10 70607-12
MFLX70607-00EA 59 EUR
MFLX70607-00 MFLX70607-10 MFLX70607-12
Plastic Hand-Operated Water and Chemical Siphon/Drum Pump
Pumput Rumpupumput
Ideal for most acid/base or ionic salt applications.
  • Portable and self-priming
These plastic hand-operated drum pumps are ideal for water or chemical siphons. All pumps are constructed with PVC barrel and hose. Water siphon model 70607-00 includes a 60” PVC outlet hose that can be cut to fit your application. Chemical siphon models 70607-10 and 70607-12 are suitable for acid and alkali transfer.