Masterflex® Adapter Fittings, Male Luer to Threaded, Straight, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX45504-84EA 38.2 EUR
MFLX45504-84 MFLX45518-82 MFLX45518-84 MFLX45518-86 MFLX45518-88
Masterflex® Adapter Fittings, Male Luer to Threaded, Straight, Avantor®
Kiinnitykset ja liittimet
Easily transition between luer fittings and UNF or NPT threads.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are male thread.