Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Nylon, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX45502-00EA 17.3 EUR
MFLX45502-00 MFLX45502-01 MFLX45502-02 MFLX45502-04 MFLX45502-05 MFLX45502-06 MFLX45502-08 MFLX45502-20 MFLX45502-16
Masterflex® Adapter Fittings, Female Luer to Hose Barb, Straight, Nylon, Avantor®
Kiinnitykset ja liittimet
Ensure your process integrity with full material traceability.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.