Masterflex® Sanitary Elbows
Toimittaja: Avantor Fluid Handling
MFLX40618-96EA 148 EUR
MFLX40618-96 MFLX40618-98 MFLX40618-88 MFLX40618-90 MFLX40618-92 MFLX40618-94
Masterflex® Sanitary Elbows
Kiinnitykset ja liittimet
These compliant, biocompatible fittings provide assurance, integrity, and cost-effective solutions.
  • Engineered to meet the needs of bioprocess workflows, single-use elbows uphold the highest standards of compliance to meet critical needs
  • Created for flawless sterility, sanitary elbows can undergo gamma irradiation and autoclaving, ensuring decontamination in all processes
  • Constructed from USP Class VI materials, sanitary elbows prioritise biocompatibility and safety, guaranteeing their suitability for sensitive bioprocess applications
  • Designed for easy