Masterflex® 1-1/2” Sanitary Tri-Clamp to 1-1/2” Sanitary Tri-Clamp, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX40330-42EA 210 EUR
MFLX40330-42 MFLX40330-43 MFLX40330-44
Masterflex® 1-1/2” Sanitary Tri-Clamp to 1-1/2” Sanitary Tri-Clamp, Avantor®
Kiinnitykset ja liittimet
Maintain sanitary conditions while easily and securely connecting process lines.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Compatible with other Tri-Clamp® connections
Tri-Clamp® unions allow for secure connection of sanitary lines. Though both materials may be autoclaved, PVDF tolerates high temperatures better than Polypropylene does and will perform more consistently after repeated autoclaving. Clamps and gaskets requiredMore Product Information