Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX40330-22EA 210 EUR
MFLX40330-22 MFLX40330-23 MFLX40330-24 MFLX40330-25 MFLX40330-26 MFLX40330-27 MFLX40330-28 MFLX40330-29 MFLX40330-30 MFLX40330-31 MFLX40330-32 MFLX40330-33 MFLX40330-34 MFLX40330-35 MFLX40330-36 MFLX40330-37 MFLX40330-38 MFLX40330-39 MFLX40330-40 MFLX40330-41
Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
Kiinnitykset ja liittimet
Maintain sanitary conditions with secure connections from sanitary Tri-Clamp to tubing.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Meet requirements for USP Class VI, FDA, REACH, and California Proposition 65
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Packaged to exacting tolerances under ISO Class 7 cleanroom conditions
  • Compatible with other Tri-Clamp® connections
The sanitary Tri-Clamp barbed adapters are ideal for use in theMore Product Information