Masterflex® Barbed Micro2 Flow Open Female Quick-Disconnect Fittings, Avantor®
Toimittaja: Avantor Fluid Handling
MFLX40262-00EA 50.2 EUR
MFLX40262-00 MFLX40262-01 MFLX40262-02 MFLX40262-03 MFLX40262-04 MFLX40262-05 MFLX40262-06 MFLX40262-07 MFLX40262-08 MFLX40262-09 MFLX40262-10 MFLX40262-11 MFLX40262-12 MFLX40262-13
Masterflex® Barbed Micro2 Flow Open Female Quick-Disconnect Fittings, Avantor®
Kiinnitykset ja liittimet
Mechanical innovations reduce risk of fluid cross-over and contamination.
  • Aseptic connection and disconnection
  • Spring-less fluid path
  • Recessed mating features
  • USP Class VI Polypropylene and PVDF housing materials
  • Elimination of accidental disconnect with safety slide release
  • A complete coupler requires a male and female fitting with same flow size (order separately). Compatible with Male Micro2 Quick-Disconnect fittings.
Masterflex® quick-disconnect fittings are an innovative and aseptic connection that allows a larger volume of fluid to pass throughMore Product Information