John Guest Acetal Push-To-Connect Male Threaded Adapter Fittings
Toimittaja: Avantor Fluid Handling
34006-67 34006-68 34006-69 34006-70 34006-71 34007-30 34007-31 34007-27 34007-28
MFLX34006-66EA 14.8 EUR
MFLX34006-66 MFLX34006-67 MFLX34006-68 MFLX34006-69 MFLX34006-70 MFLX34006-71 MFLX34007-30 MFLX34007-31 MFLX34007-27 MFLX34007-28
John Guest Acetal Push-To-Connect Male Threaded Adapter Fittings
Kiinnitykset ja liittimet
No need to buy two fitting halves.
  • Meets NSF and FDA requirements for food-grade applications
  • Quick-disconnect convenience for a wide variety of tubing material—including soft metals!
  • A single fitting offers secure connections
  • and quick disconnection
A single fitting offers secure connections and quick disconnection—no need to purchase two fitting halves. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—More Product Information