Masterflex® Direct-Read Variable-Area Flowmeters, Impact-Resistant Polycarbonate, Avantor®
Toimittaja: Avantor Fluid Handling
32900-12 32900-14 32900-16 32900-18 32900-20 32900-24 32900-26 32900-28 32900-32 32900-34 32900-38 32900-40
MFLX32900-04EA 112 EUR
MFLX32900-04 MFLX32900-08 MFLX32900-10 MFLX32900-22 MFLX32900-30 MFLX32900-36 MFLX32900-42 MFLX32900-44 MFLX32900-46 MFLX32900-48 MFLX32900-52 MFLX32900-54 MFLX32900-12 MFLX32900-14 MFLX32900-16 MFLX32900-18 MFLX32900-20 MFLX32900-24 MFLX32900-26 MFLX32900-28 MFLX32900-32 MFLX32900-34 MFLX32900-38 MFLX32900-40
Masterflex® Direct-Read Variable-Area Flowmeters, Impact-Resistant Polycarbonate, Avantor®
Virtausmittarit
Precision-adjusting valve for accurate, leak-free flow control.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Constructed with durable polycarbonate frame for rugged applications
  • Designed with graduations in English and metric for additional flexibility
Molded of high-impact, resistant polycarbonate, these direct-reading flowmeters are supplied with scales in both English and metric units for liquid and air, all with 10:1 turndown ratios. These flowmeters has been designed toMore Product Information