Masterflex® Flow Indicators for Liquids, Avantor®
Toimittaja: Avantor Fluid Handling
32779-02 32779-04 32779-06 32779-08 32779-09 32779-10
MFLX32779-00EA 157 EUR
MFLX32779-00 MFLX32880-32 MFLX32779-02 MFLX32779-04 MFLX32779-06 MFLX32779-08 MFLX32779-09 MFLX32779-10
Masterflex® Flow Indicators for Liquids, Avantor®
Virtausindikaattorit
Easily view flow through clear plastic lens.
  • Indicators feature a bright orange rotor that spins to provide visual verification
Flow indicators feature a paddle-wheel that spins at a rate proportional to the flow. Lens can be removed with a 1/4 turn for easy cleaning. Use with fluids that have a viscosity less than 34 centipoise (150 µm filter is r ecommended for protection).
Indicators/Sensors feature a Hall-effect sensor that produces a 4.5 to 24 VDC pulse output. Maximum current source output is 70 mA, and frequency output range is approximately 25More Product Information