Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Toimittaja: Avantor Fluid Handling
32776-02 32776-03
MFLX32776-00EA 208 EUR
MFLX32776-00 MFLX32776-01 MFLX32776-02 MFLX32776-03
Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Virtauksen kytkimet
Minimizes flow constriction.
  • Provides low pressure drop
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 um filter to protect your switch.
Toimitustiedot: Includes 1/4" male quick connect terminals.