Blue-White® Paddle-Wheel In-Line Flowmeters
Toimittaja: Avantor Fluid Handling
32556-02 32556-06 32556-10 32556-18 32556-24 32556-34 32556-36 32556-48 32556-52
MFLX32556-00EA 533 EUR
MFLX32556-00 MFLX32556-04 MFLX32556-08 MFLX32556-16 MFLX32556-20 MFLX32556-22 MFLX32556-32 MFLX32556-38 MFLX32556-50 MFLX32556-54 MFLX32556-58 MFLX32556-02 MFLX32556-06 MFLX32556-10 MFLX32556-18 MFLX32556-24 MFLX32556-34 MFLX32556-36 MFLX32556-48 MFLX32556-52
Blue-White® Paddle-Wheel In-Line Flowmeters
Virtausmittarit
Accurate measurement with virtually zero pressure drop.
  • Paddle wheel technology is accurate, affordable, and relatively easy to install and maintain
  • Ideally suited to open channels and partially filled pipes containing water
  • Factory calibrated for GPM or LPM
These economical in-line flowmeters are battery powered (two AAA batteries included) and thus perfect for remote locations. Enclosure is rated NEMA 4X and can be used in outdoor locations. Sensor is constructed of corrosion-resistant PVDF; in-line NPT(M) fittings are of PP.
Toimitustiedot: More Product Information