Blue-White® Direct-Read Variable-Area Flowmeters for Water, Polysulfone Housing
Toimittaja: Avantor Fluid Handling
32472-00 32472-02 32473-02 32473-04
MFLX32470-00EA 182 EUR
MFLX32470-00 MFLX32470-01 MFLX32470-02 MFLX32470-03 MFLX32470-04 MFLX32472-01 MFLX32472-03 MFLX32473-01 MFLX32472-00 MFLX32472-02 MFLX32473-02 MFLX32473-04
Blue-White® Direct-Read Variable-Area Flowmeters for Water, Polysulfone Housing
Virtausmittarit
Get better results than acrylic.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate│Use with water for laboratory and industrial applications
Choose from standard, high-capacity, or ultrapure in-line flowmeters. Standard models are designed for low flow readings—measure flow rates as low as 0.025 GPM. The high-capacity flowmeters measure flow rate as high as 130 GPM!
These large, durable flowmeters have 316 SS internal parts. New ultrapure flowmeters use only plastic wetted parts,More Product Information