Masterflex® Direct-Read Variable-Area Flowmeters with Valve, English Units, Avantor®
Toimittaja: Avantor Fluid Handling
32460-04 32460-08 32461-00 32461-08 32461-18 32466-50 32466-62
MFLX32460-00EA 126 EUR
MFLX32460-00 MFLX32460-02 MFLX32460-06 MFLX32460-10 MFLX32460-12 MFLX32460-14 MFLX32460-16 MFLX32460-22 MFLX32460-24 MFLX32461-02 MFLX32461-06 MFLX32461-07 MFLX32461-10 MFLX32461-12 MFLX32461-16 MFLX32461-22 MFLX32461-24 MFLX32461-26 MFLX32466-52 MFLX32466-60 MFLX32466-64 MFLX32460-04 MFLX32460-08 MFLX32461-00 MFLX32461-08 MFLX32461-18 MFLX32466-50 MFLX32466-62
Masterflex® Direct-Read Variable-Area Flowmeters with Valve, English Units, Avantor®
Virtausmittarit
Meter with valve provides flow control through a highly durable meter body.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium - no need to convert readings to get flow rate
  • Use with air or water for laboratory and industrial applications
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately)More Product Information